9,10-Bis(phenylethynyl)anthracene 95%
Supplier: Ambeed
|
Warning
|
A541261-100G
A541261-10G
77116-826EA
199.81
USD
77116-826
77116-828
9,10-Bis(phenylethynyl)anthracene 95%
9,10-Bis(phenylethynyl)anthracene
Caution: for Research Use Only
Formula:
C₃₀H₁₈ MW: 378.47 g/mol |
MDL Number:
MFCD00012050 CAS Number: 10075-85-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...