Indene 95% stabilised

Supplier: Ambeed
Danger

Synonyms: 1H-Indene

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A466099-25G
77116-330EA 15.45 USD
77116-330
Indene 95% stabilised
Indene

Caution: for Research Use Only

Formula: C₉H₈
MDL Number: MFCD00003777
CAS Number: 95-13-6
UN: 3295
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR