4-Amino-2,3,5,6-tetrafluorobenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2,3,5,6-tetrafluorobenzoic Acid

A457659-5G
77116-272EA 212.18 USD
77116-272
4-Amino-2,3,5,6-tetrafluorobenzoic acid 98%
4-Amino-2,3,5,6-tetrafluorobenzoic acid

Caution: for Research Use Only

Formula: C₇H₃F₄NO₂
MW: 209.1 g/mol
MDL Number: MFCD00007647
CAS Number: 944-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR