(±)-Aspartic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Aminobutanedioic acid, DL-Aspartic acid

A457463-1000G A457463-100G A457463-25G A457463-500G
77116-258EA 31.93 USD
77116-258 77116-260 77116-262 77116-264
(±)-Aspartic acid 98%
(±)-Aspartic acid

Caution: for Research Use Only

Formula: C₄H₇NO₄
MW: 133.10 g/mol
MDL Number: MFCD00063083
CAS Number: 617-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR