Dimethyl isophthalate 95%

Supplier: Ambeed
Warning

A447097-500G
77116-180EA 12.37 USD
77116-180
Dimethyl isophthalate 95%
Dimethyl isophthalate

Caution: for Research Use Only

Formula: C₁₀H₁₀O₄
MDL Number: MFCD00008433
CAS Number: 1459-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR