1,3,6-Trichloroisoquinoline 95%

Supplier: Ambeed
Danger

A389931-100MG A389931-1G A389931-250MG A389931-5G
77115-630EA 12.36 USD
77115-630 77115-632 77115-634 77594-188
1,3,6-Trichloroisoquinoline 95%
1,3,6-Trichloroisoquinoline

Caution: for Research Use Only

Formula: C₉H₄Cl₃N
MDL Number: MFCD09907620
CAS Number: 1053658-49-3
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR