1-(Difluoromethoxy)-3-nitrobenzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(Difluoromethoxy)nitrobenzene
A356068-100G
A356068-1G
A356068-25G
A356068-5G
77115-196EA
172.01
USD
77115-196
77115-198
77115-200
77115-202
1-(Difluoromethoxy)-3-nitrobenzene 98%
1-(Difluoromethoxy)-3-nitrobenzene
Caution: for Research Use Only
Formula:
C₇H₅F₂NO₃ MW: 189.12 g/mol |
MDL Number:
MFCD03407974 CAS Number: 22236-07-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...