1-(Difluoromethoxy)-3-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Difluoromethoxy)nitrobenzene

A356068-100G A356068-1G A356068-25G A356068-5G
77115-196EA 172.01 USD
77115-196 77115-198 77115-200 77115-202
1-(Difluoromethoxy)-3-nitrobenzene 98%
1-(Difluoromethoxy)-3-nitrobenzene

Caution: for Research Use Only

Formula: C₇H₅F₂NO₃
MW: 189.12 g/mol
MDL Number: MFCD03407974
CAS Number: 22236-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR