Tris(2-methylphenyl)phosphine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tris(o-tolyl)phosphine, Tri-o-tolylphosphine
A341207-1000G
A341207-100G
A341207-10G
A341207-250MG
A341207-25G
A341207-500G
A341207-5G
77115-008EA
326.51
USD
77115-008
77115-010
77115-012
77115-014
77115-016
77115-018
77115-020
Tris(2-methylphenyl)phosphine 98%
Tris(2-methylphenyl)phosphine
Caution: for Research Use Only
Formula:
C₂₁H₂₁P |
MDL Number:
MFCD00008514 CAS Number: 6163-58-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...