Lansoprazole 98%

Supplier: Ambeed
Warning

Synonyms: 2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole, 2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulfinyl)-1H-benzo[d]imidazole

A281446-100G A281446-100MG A281446-1G A281446-25G A281446-5G A281446-1MG
77114-080EA 323.42 USD
77114-080 77114-082 77114-084 77114-086 77114-088 77685-795
Lansoprazole 98%
Lansoprazole

Caution: for Research Use Only

Formula: C₁₆H₁₄F₃N₃O₂S
MW: 369.36 g/mol
MDL Number: MFCD00866873
CAS Number: 103577-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR