4'-Bromoacetophenone 95%

Supplier: Ambeed
Warning

Synonyms: p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone

A217140-1000G A217140-100G A217140-25G A217140-500G
77112-626EA 71.06 USD
77112-626 77112-628 77112-630 77112-632
4'-Bromoacetophenone 95%
4'-Bromoacetophenone

Caution: for Research Use Only

Formula: C₈H₇BrO
MDL Number: MFCD00000105
CAS Number: 99-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR