3,4-Dimethoxybenzyl alcohol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Veratryl alcohol, (3,4-Dimethoxyphenyl)methanol
A187224-100G
A187224-10G
A187224-1G
A187224-25G
A187224-500G
A187224-5G
77111-610EA
57.68
USD
77111-610
77111-612
77111-614
77111-616
77111-618
77111-620
3,4-Dimethoxybenzyl alcohol 97%
3,4-Dimethoxybenzyl alcohol
Caution: for Research Use Only
Formula:
C₉H₁₂O₃ MW: 168.19 g/mol |
MDL Number:
MFCD00004638 CAS Number: 93-03-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...