4-(Dimethylamino)cinnamaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: p-Dimethylamino-Cinnamaldehyde

A176684-100G A176684-1G A176684-250MG A176684-25G A176684-5G
77111-224EA 235.87 USD
77111-224 77111-226 77111-228 77111-230 77111-232
4-(Dimethylamino)cinnamaldehyde 98%
4-(Dimethylamino)cinnamaldehyde

Caution: for Research Use Only

Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
MDL Number: MFCD00007002
CAS Number: 6203-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR