H-Hyp(tBu)-OH 97%

Supplier: Ambeed
Warning

A166727-1G A166727-250MG A166727-25G
77110-854EA 25.75 USD
77110-854 77110-856 77110-858
H-Hyp(tBu)-OH 97%
H-Hyp(tBu)-OH

Caution: for Research Use Only

Formula: C₉H₁₇NO₃
MW: 187.239 g/mol
MDL Number: MFCD00153445
CAS Number: 79775-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR