Ethyl-3-amino-1H-pyrazole-4-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-4-carbethoxypyrazole, Ethyl 3-amino-1H-pyrazole-4-carboxylate

A164006-100G A164006-10G A164006-1G A164006-25G A164006-500G A164006-5G
77110-774EA 47.39 USD
77110-774 77110-776 77110-778 77110-780 77110-782 77110-784
Ethyl-3-amino-1H-pyrazole-4-carboxylate 95%
Ethyl 3-amino-1H-pyrazole-4-carboxylate

Caution: for Research Use Only

Formula: C₆H₉N₃O₂
MDL Number: MFCD00005238
CAS Number: 6994-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR