DL-Methionine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-4-(methylthio)butanoic acid, (±)-2-Amino-4-(methylthio)butyric acid, Methioninum racemicum, Methionine racematic, H-DL-Met-OH, (±)-Methionine, (±)-2-Amino-4-(methylthio)butanoic acid
A150589-1000G
A150589-100G
A150589-25G
A150589-500G
77110-278EA
29.87
USD
77110-278
77110-280
77110-282
77110-284
DL-Methionine 98%
DL-Methionine
Caution: for Research Use Only
Formula:
C₅H₁₁NO₂S |
MDL Number:
MFCD00063096 CAS Number: 59-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...