Tadalafil 98%

Supplier: Ambeed
Warning

Synonyms: (6R,12aR)-6-(Benzo[d][1,3]dioxol-5-yl)-2-methyl-2,3,12,12a-tetrahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4(6H,7H)-dione

A147960-1000G A147960-100G A147960-100MG A147960-10G A147960-500G A147960-1G A147960-5G A147960-25G
103803-854EA 9 USD
103803-854 103841-970 77110-114 77110-116 77110-118 77110-120 77110-122 77299-618 77299-620 77317-394
Tadalafil 98%
Tadalafil

Caution: for Research Use Only

Formula: C₂₂H₁₉N₃O₄
Boiling Pt: 679.1 °C (760 mm Hg)
Melting Pt: 298…300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07771966
CAS Number: 171596-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR