DL-Lysine monohydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(±)-2,6-Diaminocaproic acid monohydrochloride, H-DL-Lys-OH · HCl, H-DL-Lys-OH.HCl, (±)-2,6-Diaminohexanoic acid monohydrochloride, (±)-Lysine monohydrochloride
A142323-100G
A142323-10G
A142323-500G
A142323-5G
A142323-25G
77109-864EA
49.45
USD
77109-864
77109-866
77109-868
77109-870
77316-358
DL-Lysine monohydrochloride 95%
DL-Lysine monohydrochloride
Caution: for Research Use Only
Formula:
C₆H₁₅ClN₂O₂ |
MDL Number:
MFCD00064563 CAS Number: 70-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...