2-Acetamido-5-bromopyridine 98%

Supplier: Ambeed
Warning

A139145-25G
77109-732EA 64.89 USD
77109-732
2-Acetamido-5-bromopyridine 98%
2-Acetamido-5-bromopyridine

Caution: for Research Use Only

Formula: C₇H₇BrN₂O
MDL Number: MFCD00468968
CAS Number: 7169-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR