4-Hydroxyphenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid
A137634-1000G
A137634-100G
A137634-10G
A137634-500G
A137634-25G
A137634-5G
A137634-1G
103802-478EA
5
USD
103802-478
103833-466
77109-626
77109-628
77109-630
77109-632
77298-914
77298-916
77315-450
4-Hydroxyphenylboronic acid 97%
4-Hydroxyphenylboronic acid
Caution: for Research Use Only
Formula:
C₆H₇BO₃ MW: 137.93 g/mol Melting Pt: >230 °C Flash Pt: 166.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074628 CAS Number: 71597-85-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...