Methyl-4-hydroxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate

A137460-1000G A137460-100G A137460-25G A137460-500G
77109-612EA 36.04 USD
77109-612 77109-614 77109-616 77109-618
Methyl-4-hydroxybenzoate 98%
Methyl 4-hydroxybenzoate

Caution: for Research Use Only

Formula: HOC₆H₄CO₂CH₃
MW: 152.15 g/mol
MDL Number: MFCD00002352
CAS Number: 99-76-3
Merck Index: 13,06129

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR