L(+)-Aspartic acid 98%

Supplier: Ambeed
Warning

A136286-1000G A136286-100G A136286-25G A136286-500G
77109-558EA 23.69 USD
77109-558 77109-560 77109-562 77109-564
L(+)-Aspartic acid 98%
L(+)-Aspartic acid

Caution: for Research Use Only

Formula: C₄H₇NO₄
MDL Number: MFCD00002616
CAS Number: 56-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR