2-Chloro-5-fluorobenzoic acid 95%

Supplier: Ambeed
Warning

A127468-10G A127468-25G A127468-5G
77108-988EA 11.33 USD
77108-988 77108-990 77108-992 77108-994
2-Chloro-5-fluorobenzoic acid 95%
2-Chloro-5-fluorobenzoic acid

Caution: for Research Use Only

Formula: C₇H₄ClFO₂
MDL Number: MFCD00156967
CAS Number: 2252-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR