Ethyl nicotinate 98%
Supplier: Ambeed
|
Warning
|
A107966-100G
A107966-10G
A107966-25G
A107966-500G
A107966-1000G
103807-986EA
51
USD
103807-986
77107-786
77107-788
77107-790
77107-792
77309-890
Ethyl nicotinate 98%
Ethyl nicotinate
Caution: for Research Use Only
Formula:
C₈H₉NO₂ Boiling Pt: 224 °C (760 mm Hg) Melting Pt: 8…10 °C Flash Pt: 93.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006389 CAS Number: 614-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...