1,2,3-Tri-O-acetyl-5-deoxy-β-D-ribofuranose 97%

Supplier: Ambeed
Warning

A105837-100G A105837-10G A105837-25G A105837-500G A105837-5G
103822-698EA 6 USD
103822-698 77107-600 77107-602 77107-604 77107-606 77107-608 77283-404
1,2,3-Tri-O-acetyl-5-deoxy-β-D-ribofuranose 97%
1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose

Caution: for Research Use Only

Formula: C₁₁H₁₆O₇
Boiling Pt: 315.3 °C (760 mm Hg)
Melting Pt: 63…64 °C
Flash Pt: 136 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08458459
CAS Number: 62211-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR