1,3-Dibromobenzene 98%
Supplier: Ambeed
|
Warning
|
A103634-100G
A103634-10G
A103634-25G
A103634-500G
A103634-1000G
A103634-5G
103807-872EA
173
USD
103807-872
103822-296
77107-356
77107-358
77107-360
77107-362
77309-166
77309-168
1,3-Dibromobenzene 98%
1,3-Dibromobenzene
Caution: for Research Use Only
Formula:
C₆H₄Br₂ Boiling Pt: 218…219 °C (760 mm Hg) Melting Pt: 7 °C Flash Pt: 93.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000078 CAS Number: 108-36-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...