1,3-Dibromobenzene 98%

Supplier: Ambeed
Warning

A103634-100G A103634-10G A103634-25G A103634-500G A103634-1000G A103634-5G
103807-872EA 173 USD
103807-872 103822-296 77107-356 77107-358 77107-360 77107-362 77309-166 77309-168
1,3-Dibromobenzene 98%
1,3-Dibromobenzene

Caution: for Research Use Only

Formula: C₆H₄Br₂
Boiling Pt: 218…219 °C (760 mm Hg)
Melting Pt: 7 °C
Flash Pt: 93.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00000078
CAS Number: 108-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR