2-Methoxy-6-quinolineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

A779557-100MG A779557-1G A779557-250MG
77105-432EA 37 USD
77105-432 77105-434 77105-436
2-Methoxy-6-quinolineboronic acid pinacol ester 98%
2-Methoxy-6-quinolineboronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₆H₂₀BNO₃
MDL Number: MFCD18383956
CAS Number: 1201644-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR