Gemfibrozil 98%

Supplier: Ambeed
Warning

Synonyms: 5-(2,5-Dimethylphenoxy)-2,2-dimethylpentanoic acid, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)pentanoic acid, CI-719, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)valeric acid

A640007-100G A640007-100MG A640007-10G A640007-5G A640007-25G A640007-500G
77104-398EA 80 USD
77104-398 77104-400 77104-402 77104-404 77569-226 77686-585
Gemfibrozil 98%
Gemfibrozil

Caution: For research use only.

Formula: C₁₅H₂₂O₃
MW: 250.34
Boiling Pt: 395
MDL Number: MFCD00079335
CAS Number: 25812-30-0
Merck Index: 12,04394

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR