Daidzin 98+%

Supplier: Ambeed
Warning

Synonyms: 7-(beta-D-Glucopyranosyloxy)-4'-hydroxyisoflavone, 3-(4-Hydroxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

A590254-100MG A590254-10MG A590254-1G A590254-1MG A590254-250MG
77103-888EA 64 USD
77103-888 77103-890 77103-892 77103-894 77103-896
Daidzin 98+%
Daidzin

Caution: for Research Use Only

Formula: C₂₁H₂₀O₉
MW: 416.38 g/mol
MDL Number: MFCD00017466
CAS Number: 552-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR