2-Chlorothioanisole 97%

Supplier: Ambeed
Warning

A507495-25G A507495-5G A507495-100G
77103-064EA 41 USD
77103-064 77103-066 77901-576
2-Chlorothioanisole 97%
2-Chlorothioanisole

Caution: for Research Use Only

Formula: C₇H₇ClS
MDL Number: MFCD00040124
CAS Number: 17733-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR