Hesperetin 97%

Supplier: Ambeed
Warning

Synonyms: 3',5,7-Trihydroxy-4'-methoxyflavanone, 4'-Methoxy-3',5,7-trihydroxyflavanone, (S)-5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)chroman-4-one, (±)-Hesperetin

A238234-100G A238234-10G A238234-500G A238234-50MG A238234-5G A238234-1G A238234-25G
103811-212EA 6 USD
103811-212 77098-402 77098-404 77098-406 77098-408 77098-410 77304-100 77304-102
Hesperetin 97%
Hesperetin

Caution: for Research Use Only

Formula: C₁₆H₁₄O₆
MW: 302.30 g/mol
Boiling Pt: 586.2 °C (760 mm Hg)
Melting Pt: 227…232 °C
Flash Pt: 223 °C
Storage Temperature: Ambient
MDL Number: MFCD00075646
CAS Number: 520-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR