Fmoc-Dap(Alloc)-OH 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Dap(Aloc)-OH, (S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid

A210001-1G A210001-250MG A210001-5G A210001-10G A210001-25G A210001-100G
77097-638EA 23 USD
77097-638 77097-640 77097-642 77568-024 77612-060 77685-545
Fmoc-Dap(Alloc)-OH 95%
Fmoc-Dap(Alloc)-OH

Caution: For research use only.

Formula: C₂₂H₂₂N₂O₆
MDL Number: MFCD00273468
CAS Number: 188970-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR