2-Formyl-4-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methylpyridine-2-carboxaldehyde, 4-Methylpicolinaldehyde

A166216-100G A166216-1G A166216-250MG A166216-25G A166216-5G
77096-322EA 481 USD
77096-322 77096-324 77096-326 77096-328 77096-330
2-Formyl-4-picoline 98%
2-Formyl-4-picoline

Caution: for Research Use Only

Formula: C₇H₇NO
MDL Number: MFCD07367933
CAS Number: 53547-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR