5,6-Dichlorobenzimidazole Hydrochloride 95%

Supplier: Ambeed
Warning

A161668-250MG
77096-184EA 10 USD
77096-184
5,6-Dichlorobenzimidazole Hydrochloride 95%
5,6-Dichlorobenzimidazole Hydrochloride

Caution: for Research Use Only

Formula: C₇H₄Cl₂N₂
MDL Number: MFCD16810292
CAS Number: 1087737-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR