H-D-Tyr(tBu)-OH 97%

Supplier: Ambeed
Warning

A142265-1G A142265-250MG A142265-5G
77095-468EA 45 USD
77095-468 77095-470 77900-609
H-D-Tyr(tBu)-OH 97%
H-D-Tyr(tBu)-OH

Caution: for Research Use Only

Formula: C₁₃H₁₉NO₃
MW: 237.299 g/mol
MDL Number: MFCD00077112
CAS Number: 186698-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR