FK-866 ≥98%

Supplier: Ambeed
Warning

Synonyms: (E)-N-(4-(1-Benzoylpiperidin-4-yl)butyl)-3-(pyridin-3-yl)acrylamide, (E)-Daporinad

A129216-10MG A129216-1MG A129216-5MG A129216-50MG A129216-100MG A129216-1G A129216-250MG
77094-910EA 56 USD
77094-910 77094-912 77094-914 77611-146 77685-119 77685-120 77685-121
FK-866 ≥98%
FK-866

Caution: for Research Use Only

Formula: C₂₄H₂₉N₃O₂
MDL Number: MFCD10565943
CAS Number: 658084-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR