1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one 98%

Supplier: Ambeed
Danger

A124580-100G A124580-10G A124580-1G A124580-25G A124580-5G
77094-688EA 59 USD
77094-688 77094-690 77094-692 77094-694 77094-696
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one 98%
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one

Caution: for Research Use Only

Formula: C₅H₄N₄O
MDL Number: MFCD00599413
CAS Number: 315-30-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR