1,4-Dioxaspiro[4.5]decan-8-one 98%

Supplier: Ambeed
Warning

Synonyms: 1,4-Cyclohexanedione monoethylene acetal, 1,4-Cyclohexanedione monoethylene ketal

A123676-100G A123676-10G A123676-1G A123676-25G A123676-500G A123676-5G
103808-280EA 303 USD
103808-280 77094-604 77094-606 77094-608 77094-610 77094-612 77094-614
1,4-Dioxaspiro[4.5]decan-8-one 98%
1,4-Dioxaspiro[4.5]decan-8-one

Caution: for Research Use Only

Formula: C₈H₁₂O₃
MW: 156.18 g/mol
Melting Pt: 70…73 °C
Flash Pt: 106.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00010214
CAS Number: 4746-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR