Imperatorin 97%

Supplier: Ambeed
Warning

Synonyms: 9-((3-Methylbut-2-en-1-yl)oxy)-7H-furo[3,2-g]chromen-7-one

A118402-10MG A118402-1MG A118402-25MG A118402-5MG A118402-1G A118402-250MG
77094-342EA 21 USD
77094-342 77094-344 77094-346 77094-348 77610-970 77685-045
Imperatorin 97%
Imperatorin

Caution: for Research Use Only

Formula: C₁₆H₁₄O₄
MDL Number: MFCD00016881
CAS Number: 482-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR