2-Chloronicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-pyridinecarboxylic acid, 2-Chloropyridine-3-carboxylic acid

A103202-1000G A103202-100G A103202-25G A103202-500G
77093-604EA 55 USD
77093-604 77093-606 77093-608 77093-610
2-Chloronicotinic acid 98%
2-Chloronicotinic acid

Caution: for Research Use Only

Formula: C₆H₄ClNO₂
MDL Number: MFCD00006236
CAS Number: 2942-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR