2-Chloronicotinic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-3-pyridinecarboxylic acid, 2-Chloropyridine-3-carboxylic acid
A103202-1000G
A103202-100G
A103202-25G
A103202-500G
77093-604EA
55
USD
77093-604
77093-606
77093-608
77093-610
2-Chloronicotinic acid 98%
2-Chloronicotinic acid
Caution: for Research Use Only
Formula:
C₆H₄ClNO₂ |
MDL Number:
MFCD00006236 CAS Number: 2942-59-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...