High Pressure Controller
Supplier: Lighthouse Worldwide
402965266-1
77093-406EA 6077.24 USD
77093-406
High Pressure Controller
Samplers Gas Samplers
The High Pressure Controller is used in conjuction with the Solair 1100 to sample high pressure gasses.
  • Supports 0.1 µm at 1 CFM
  • May be used with any inert gas
  • Simple installation, easy setup
  • Front-mounted flow gauge
  • Intuitive user control
  • Quiet operation
  • Fast purge and cleanup
  • VCR fittings internally and externally
  • Stainless steel construction
Designed using the latest technology in high pressure gas components, the HPC 1100 allows attachment of Lighthouse 0.1 µm particle counters to high pressure (30 to 150 psi) inert gas systems to provide monitoring capability for process gas contamination.
The HPC 1100’s exceptionally low audible noise and small footprint allow installation in areas where diffusers were thought impractical. The HPC 1100 zero counts in less than 5 minutes and does not induce spurious or random 0.1 µm counts due to oscillations as other designs can.
The HPC 1100’s stainless steel design is rugged, reliable, requires almost no routine maintenance, and never requires calibration. During normal operation, no inert gas is wasted. 100% of the gas supplied is used for the sampling process.