Liquid Batch Sampling System, LS-20
Supplier: Lighthouse Worldwide
403006201-1
77093-404EA 27311.88 USD
77093-404
Liquid Batch Sampling System, LS-20
Particle Counters
Lighthouse’s Liquid Batch Samplers are designed to be user-friendly and intuitive for utilization. Most importantly, it is designed to be compliant to industry standards.
  • 1.0 to 50 µm or 1.0 to 200 µm size range
  • Integrated 'light obscuration' particle counter software designed for 21 CFR part 11
  • International regulatory EP, JP reports
  • USP 788 reports
  • Sample volumes from 1 to 1000 ml
  • Preconfigured USP 788 recipes
  • Precise syringe pump flow control
  • Stainless steel enclosure
  • Built-in magnetic stirrer
  • Automated sample handling maximizes repeatability
  • Compact footprint