Liquid Batch Sampling System, LS-60
Supplier: Lighthouse Worldwide
403006200-1
77093-396EA 48232.97 USD
77093-396
Liquid Batch Sampling System, LS-60
Particle Counters
Lighthouse’s Liquid Batch Samplers are designed to be user-friendly and intuitive for utilization. Most importantly, it is designed to be compliant to industry standards.
  • 0.1 to 100 µm size ranges available
  • Integrated particle counter
  • 8 channel sizes
  • Windows-based control software
  • Sample volumes from 1 ml to 1 L
  • Syringe size options: 10 and 25 ml
  • Precise syringe pump flow control
  • Stainless steel enclosure
  • Built-in magnetic stirrer
  • Maximizes sample repeatability
  • Small footprint
  • RS485 Modbus
  • Designed for reliability