Particle Counter, ApexZ50
Supplier: Lighthouse Worldwide
402999471-2
77093-392EA 12793.65 USD
77093-392
Particle Counter, ApexZ50
Particle Counters
The ApexZ50 contains the most complete system of self-diagnostics in the industry, an integrated alarm indicator in the handle, an 11 hour battery life with hot-swappable smart batteries, and both WiFi and USB communication.
  • Particle size range: 0.5 - 25 μm
  • Flow rate: 100 LPM
  • Bi-directional integration into LIMS EM Systems
  • Sealed chemically compatible enclosure
  • Advanced self-diagnostics
  • Integrated status lights
  • Hot-swappable smart batteries
  • Wi-Fi, Ethernet, and USB data transfer options
  • 21 CFR Part 11 compliant
  • Customizable data reports
  • Built-in cleanroom certification reports
  • 2-year warranty
Certifications: CE. FCC.