Pentafluorophenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,3,4,5,6-Pentafluorobenzeneboronic acid, Dihydroxy(pentafluorophenyl)borane, (Pentafluorophenyl)metaboric acid, Pentafluorobenzeneboronic acid
A268671-1G
A268671-25G
A268671-10G
A268671-5G
A268671-100G
103812-928EA
18
USD
103812-928
103852-830
77092-588
77092-590
77153-598
77305-012
77900-932
Pentafluorophenylboronic acid 97%
Pentafluorophenylboronic acid
Caution: for Research Use Only
Formula:
C₆H₂BF₅O₂ Boiling Pt: 244 °C (760 mm Hg) Melting Pt: 176…179 °C Flash Pt: 101.4 °C Storage Temperature: Freezer |
MDL Number:
MFCD01074663 CAS Number: 1582-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...