Chloro(1,5-cyclooctadiene)rhodium(I) dimer 98%

Supplier: Ambeed
Warning

Synonyms: (1,5-Cyclooctadiene)rhodium chloride dimer

A254450-100MG A254450-250MG A254450-1G A254450-5G
103812-140EA 290 USD
103812-140 103851-994 76692-588 76692-590 77091-436 77091-438
Chloro(1,5-cyclooctadiene)rhodium(I) dimer 98%
Chloro(1,5-cyclooctadiene)rhodium (I) dimer

Caution: for Research Use Only

Formula: C₁₆H₂₄Cl₂Rh₂
MW: 493.08 g/mol
Melting Pt: 243 °C
Storage Temperature: Ambient
MDL Number: MFCD00012415
CAS Number: 12092-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR