2-(Dicyclohexylphosphino)biphenyl 98%

Supplier: Ambeed
Warning

Synonyms: (2-Biphenyl)dicyclohexylphosphine

A249737-25G A249737-5G A249737-100G A249737-10G A249737-500G A249737-1G
103811-888EA 5 USD
103811-888 103851-726 77091-066 77091-068 77238-242 77238-244 77238-246 77304-498
2-(Dicyclohexylphosphino)biphenyl 98%
2-(Dicyclohexylphosphino)biphenyl

Caution: for Research Use Only

Formula: C₂₄H₃₁P
MW: 350.48 g/mol
Flash Pt: 271.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01862441
CAS Number: 247940-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR