(S)-(+)-β-Aminophenethyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol

A238684-100G A238684-10G A238684-1G A238684-500G A238684-1000G A238684-25G A238684-5G
103809-686EA 191 USD
103809-686 103851-030 76771-344 76771-346 76771-348 76771-350 77090-156 77090-158 77090-160
(S)-(+)-β-Aminophenethyl alcohol 98%
(S)-(+)-β-Aminophenethyl alcohol

Caution: for Research Use Only

Formula: C₈H₁₁NO
Flash Pt: 125.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00064404
CAS Number: 20989-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR